Orders LinkContactUsLinkComponent
Polypropylene Beaker Set
Polypropylene Beaker Set
50 ml, 100 ml, 250 ml, 500 ml, 1000 ml, Set of 5
Catalog # 470005-162
Supplier:   Dynalon
Supplier #:  222195
Polypropylene Beaker Set
Catalog # 470005-162
Supplier:  Dynalon
Polypropylene Beaker Set
Polypropylene Beaker Set
50 ml, 100 ml, 250 ml, 500 ml, 1000 ml, Set of 5
Price
$57.00
Log in for availability
Each Kit of 5
Product Details & Documents
Unbreakable polypropylene.
  • Autoclavable
  • Chemical-resistant
These beaker sets autoclavable up to 145 °C and chemical resistant. All feature pour spouts and metric graduation scales.
Specifications
  • Capacity:
    50 ml, 100 ml, 250 ml, 500 ml, 1000 ml
  • Description:
    Set of 5