Plane Mirrors
Plane Mirrors, Size, mm: 152 x 38 x 2.5
Catalog # 470111-288
Supplier: Abrisa-USPG Industrial Glass
Supplier #: 2-393
Jump to:
Product Details & Documents
All Types and Sizes for All Optical Applications
- Front surface silvering eliminates double reflections
- All produce virtual images
These plane mirrors are rectangular. They are available in glass or shatterproof Plexiglas. Back surface silvered mirrors are less expensive, and ideal for general use. The silvered back surface is painted for durability. Front surface silvering eliminates double reflections at the unsilvered surface, giving a brighter, sharper image. However, these mirrors are more easily damaged by scratching.
Specifications
- Description:Plane Mirrors, Size, mm: 152 x 38 x 2.5