Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Environmentally Friendly Thermometers

Environmentally Friendly Thermometers, 1 °C, Total Immersion, −20...+110 °C, 12"
Catalog # 470019-644
Supplier:   Thermco
 :  B110CYBLK
Environmentally Friendly Thermometers
Catalog # 470019-644
Supplier:  Thermco
Environmentally Friendly Thermometers
Environmentally Friendly Thermometers

Environmentally Friendly Thermometers, 1 °C, Total Immersion, −20...+110 °C, 12"

Price
$21.00
Log in for availability
Each Box of 10
Product Details & Documents

Glass thermometers with partial or total immersion.

Specifications
  • Description:
    Environmentally Friendly Thermometers, 1 °C, Total Immersion
  • Length:
    12"
  • Range:
    −20...+110 °C
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.