Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Model Plus: Cells

Model Plus: Cells

Model Plus: The Animal Cell, 28 cm, 28 cm, 37 cm
Catalog # 470016-890
Supplier:   SI Manufacturing
Supplier #:  69072
Model Plus: Cells
Catalog # 470016-890
Supplier:  SI Manufacturing
Model Plus: Cells
Model Plus: Cells

Model Plus: The Animal Cell, 28 cm, 28 cm, 37 cm

Price
$92.99
Log in for availability
Each
Product Details & Documents

Learn Cell Structure With Hands-On Foam Pieces

  • Easy To Manipulate Pieces
  • Guides Allow Students To Build And Learn At Their Own Pace
  • Designed For Grades 1-8

Simple for elementary level students yet comprehensive enough for middle school, these colorful teaching models Were designed specifically for hands-on use. Each model is made of resilient, non-toxic EVA foam to withstand years of active investigation. A thorough, detailed teacher’s guide includes inquiry-based lab activities, reproducible worksheets, complete background information, cross-curricular extension activities, assessment ideas and more. Topics covered include cell structure and function, osmosis, eukaryotes, bacteria and more.

Specifications
  • Description:
    Model Plus: The Animal Cell
  • Height:
    37 cm
  • Length:
    28 cm
  • Width:
    28 cm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.