Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Somso® Functional Joints

Functional Ankle and Foot Joints, 1, 18 cm, 28 cm, 28 cm
  470015-620
 :  NS 54
Somso® Functional Joints
  470015-620
Somso® Functional  Joints
Somso® Functional Joints

Functional Ankle and Foot Joints, 1, 18 cm, 28 cm, 28 cm

 
$1,090.00
   
Each
 

A unique manufacturing process makes these models exceptionally lifelike. All feature a durable, yet flexible, rubberized material applied over molded plastic bone structures allowing for flexion and extension, inner and outer rotation, adduction and abduction, and other realistic movements of the joints.

  • Joints move in a similar range of motion as normal bones
  • Each joint is supplied on its own base
  • 5 year manufacturers warranty

Models include a teacher's guide.

 
  • Description:
    Functional Ankle and Foot Joints
  • Height:
    28 cm
  • Length:
    28 cm
  • Width:
    18 cm
  • No. of parts:
    1