Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Fermentation Tubes

Fermentation Tubes

Fermentation Tube, Ungraduated, 10 mL, with Foot
Catalog # 470002-074
Supplier:   GSC International
Supplier #:  601-2
Fermentation Tubes
Catalog # 470002-074
Supplier:  GSC International
Fermentation Tubes
Fermentation Tubes

Fermentation Tube, Ungraduated, 10 mL, with Foot

Price
$17.90
Log in for availability
Each
Product Details & Documents

Verify gas production in fermentation exercises, gas produced by yeast or other organisms rises into a 5 or 10 ml evolution tube.

  • Ungraduated with foot
Specifications
  • Description:
    Fermentation Tube, Ungraduated, 10 mL, with Foot
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.