Orders LinkContactUsLinkComponent
Differential Quick Stain, Electron Microscopy Sciences
Differential Quick Stain, Electron Microscopy Sciences
Catalog # 102773-658
Differential Quick Stain, Electron Microscopy Sciences
Catalog # 102773-658
Supplier:  
CAS Number:  
Differential Quick Stain, Electron Microscopy Sciences
CAS Number:  
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This kit is a Modified Giemsa Stain kit for quick turnaround results. It may be used for rapid blood smears for differential assessment, as well as for the detection of H. Pylori microorganisms.
  • The stain offers results in 15 seconds
  • Comes with a fixative for air dried cell suspensions and or touch preparation slides
Used for fine needle aspirations, frozen sections, blood smears, cytological specimens, cytopreps, microorganism detection and bone marrow biopsies.
Certifications: Certificates of analysis are available.
Ordering information: The Kit comes complete with 3 solutions and is available in 3 sizes.