Orders LinkContactUsLinkComponent
Differential Quick Stain, Electron Microscopy Sciences
Differential Quick Stain, Electron Microscopy Sciences
Differential Quick Stain Kit, 250 mL
Catalog # 102773-654
Supplier #:  26096-25
Differential Quick Stain, Electron Microscopy Sciences
Catalog # 102773-654
CAS Number:  
Differential Quick Stain, Electron Microscopy Sciences
Differential Quick Stain Kit, 250 mL
CAS Number:  
Price
$324.43
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
This kit is a Modified Giemsa Stain kit for quick turnaround results. It may be used for rapid blood smears for differential assessment, as well as for the detection of H. Pylori microorganisms.
  • The stain offers results in 15 seconds
  • Comes with a fixative for air dried cell suspensions and or touch preparation slides
Used for fine needle aspirations, frozen sections, blood smears, cytological specimens, cytopreps, microorganism detection and bone marrow biopsies.
Certifications: Certificates of analysis are available.
Ordering information: The Kit comes complete with 3 solutions and is available in 3 sizes.
Specifications
Specifications
  • Description:
    Differential Quick Stain Kit
  • Size:
    250 mL