Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Certipur® pH Buffer Solutions, MilliporeSigma

Boric acid/potassium chloride/sodium hydroxide solution, Borate, 25 °C, Sachet, 30×30 ml, 9.00, Colorless
Catalog # EM1.99003.0001
Supplier:   Avantor
Certipur® pH Buffer Solutions, MilliporeSigma
Catalog # EM1.99003.0001
Supplier:  Avantor
CAS Number:  
Certipur® pH Buffer Solutions, MilliporeSigma
Certipur® pH Buffer Solutions, MilliporeSigma

Boric acid/potassium chloride/sodium hydroxide solution, Borate, 25 °C, Sachet, 30×30 ml, 9.00, Colorless

CAS Number:  
Price
$259.33
Log in for availability
Each Case of 16
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Specifications

Specifications

  • Pack type:
    Sachet
  • Color:
    Colorless
  • Description:
    Boric acid/potassium chloride/sodium hydroxide solution
  • Size:
    30×30 ml
  • pH buffer type:
    Borate
  • Reference temperature:
    25 °C
  • pH:
    9.00

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.