Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Supelco® Certipur® pH Buffer Solutions

Boric acid/potassium chloride/sodium hydroxide, Borate, 20 °C, Bag in box, Titripac®, 4 L, 9.00, Colorless
Catalog # EM1.09461.4000
   Sustainable Packaging
Supplier:   MilliporeSigma
 :  1.09461.4000
Supelco® Certipur® pH Buffer Solutions
Catalog # EM1.09461.4000
Supplier:  MilliporeSigma
   Sustainable Packaging
CAS Number:  
Supelco® Certipur® pH Buffer Solutions
Supelco® Certipur® pH Buffer Solutions

Boric acid/potassium chloride/sodium hydroxide, Borate, 20 °C, Bag in box, Titripac®, 4 L, 9.00, Colorless

CAS Number:  
Price
$207.00
Log in for availability
Each (4l) Case of 4 (4l)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Specifications

Specifications

  • Pack type:
    Bag in box, Titripac®
  • Color:
    Colorless
  • Description:
    Boric acid/potassium chloride/sodium hydroxide
  • Size:
    4 L
  • pH buffer type:
    Borate
  • Reference temperature:
    20 °C
  • pH:
    9.00
  • Cat. No.:
    EM1.09461.4000
  • Environmentally Preferable:
    Sustainable Packaging

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.