Certipur® pH Buffer Solutions, MilliporeSigma
Di-sodium hydrogen phosphate/potassium di-hydrogen phosphate, Phosphate, 20 °C, Bag in box, Titripac®, 10 L, 7.00, Colorless
Supplier: MilliporeSigma
: 1094399010
Certipur® pH Buffer Solutions, MilliporeSigma
Catalog # EM1.09439.9010
Supplier: MilliporeSigma
CAS Number:
Jump to:
Product Details & Documents
Traceable to SRM from NIST and PTB.
Certificates
Search by Lot Number1.07827.1000_08072012 1.09400.0500_08162012 1.09400.4000_08162012 1.09400.9010_08162012 1.09407.0500_08022012
More
Specifications
Specifications
- Pack type:Bag in box, Titripac®
- Color:Colorless
- Description:Di-sodium hydrogen phosphate/potassium di-hydrogen phosphate
- Size:10 L
- pH buffer type:Phosphate
- Reference temperature:20 °C
- pH:7.00
- Cat. No.:EM1.09439.9010
- Environmentally Preferable:Sustainable Packaging
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



