Supelco® Certipur® pH Buffer Solutions
Citric acid/sodium hydroxide/hydrogen chloride, Citrate, 20 °C, Bag in box, Titripac®, 10 L, 4.01, Colorless
Supplier: MilliporeSigma
: 1094359010
Supelco® Certipur® pH Buffer Solutions
Catalog # EM1.09435.9010
Supplier: MilliporeSigma
CAS Number:
Jump to:
Product Details & Documents
Traceable to SRM from NIST and PTB.
Documents
Certificates
Search by Lot Number1.07827.1000_08072012 1.09400.0500_08162012 1.09400.4000_08162012 1.09400.9010_08162012 1.09407.0500_08022012
SDS
More
Avantor Focus Production Volume 3 Issue 1 AYN4_Analytical_Chemistry_-Electrochemistry EMD Millipore Certipur Reference Materials Catalog 051795W EMD Millipore Sustainable Products Brochure Merck Buffer Calibration Standards for pH Measurements
Product Literature
More
Certipur® Ready-to-Use pH Calibration Buffer Solutions
Additional Product Details
TDS Certified Reference Materials
Datasheet
Specifications
Specifications
- Pack type:Bag in box, Titripac®
- Color:Colorless
- Description:Citric acid/sodium hydroxide/hydrogen chloride
- Size:10 L
- pH buffer type:Citrate
- Reference temperature:20 °C
- pH:4.01
- Cat. No.:EM1.09435.9010
- Environmentally Preferable:Sustainable Packaging
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



