BCSA Base, CRITERION™, Hardy Diagnostics
122.4 g, 2 L, Zip-pouch to make 2 L, Mylar®
Supplier: Hardy Diagnostics
Supplier #: C8880
BCSA Base, CRITERION™, Hardy Diagnostics
Catalog # 89406-812
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Jump to:
Product Details & Documents
CRITERION™ BCSA Base, used for the selective isolation of Burkholderia (Pseudomonas) cepacia
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Antibiotics Polymixin B, Gentamicin, and Vancomycin required. See product label for details. Dehydratedculture media with engineered container.
Certifications: BSE Free
Documents
Certificates
Search by Lot NumberHardy Diagnostics Criterion Dehydrated Culture Media
Product Literature
Specifications
Specifications
- Pack type:Zip-pouch to make 2 L, Mylar®
- Size:122.4 g
- Final volume:2 L
- Cat. No.:89406-812
- Storage Temperature:–2-30°C
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



