CRITERION™ Phosphate Buffer, pH 7.2
500 g, Bottle, wide mouth
Supplier: Hardy Diagnostics
Supplier #: C8331
CRITERION™ Phosphate Buffer, pH 7.2
Catalog # 89406-750
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Jump to:
Product Details & Documents
CRITERION™ Agar, bacteriological grade is used as a solidifying agent in the preparation of solid and semi-solid culture media.
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- Increased solubility - dissolves quickly
- Custom and bulk products available - to your specifications
- BSE-free
Documents
Specifications
Specifications
- Pack type:Bottle, wide mouth
- Size:500 g
- Application:Breweries, wineries and distilleries, cannabis QA testing, clinical, cosmetics, environmental, food safety, compounding pharmacy, pharmaceutical manufacturing
- Cat. No.:89406-750
- Storage Temperature:2...30 °C



