Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Indole Nitrate Medium, CRITERION™, Hardy Diagnostics

122.4 g, 2 L, Zip-pouch to make 2 L, Mylar®
  89405-496
Product available on GSA Advantage®
 :  C5870
Indole Nitrate Medium, CRITERION™, Hardy Diagnostics
  89405-496
Product available on GSA Advantage®
 :  
Indole Nitrate Medium, CRITERION™, Hardy Diagnostics
Indole Nitrate Medium, CRITERION™, Hardy Diagnostics

122.4 g, 2 L, Zip-pouch to make 2 L, Mylar®

 :  
 
$72.60
   
Each
 
 

CRITERION™ Indole Nitrate Medium for the detection of indole production and nitrate reduction by microorganisms

  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications

Dehydratedculture media with engineered container.

Certifications: BSE Free

 

 

  • Pack type:
    Zip-pouch to make 2 L, Mylar®
  • Size:
    122.4 g
  • Final volume:
    2 L
  • Cat. No.:
    89405-496
  • Storage Temperature:
    –2-30°C

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.