Eugonic Agar, CRITERION™, Hardy Diagnostics
500 g, Bottle, wide mouth
Supplier: Hardy Diagnostics
Supplier #: C5721
Eugonic Agar, CRITERION™, Hardy Diagnostics
Catalog # 89405-438
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Jump to:
Product Details & Documents
CRITERION™ Eugonic Agar for the cultivation of microorganisms
Dehydratedculture media with engineered container.
Documents
Certificates
Search by Lot NumberHardy Diagnostics Criterion Dehydrated Culture Media
Product Literature
Specifications
Specifications
- Pack type:Bottle, wide mouth
- Size:500 g
- Cat. No.:89405-438
- Storage Temperature:–2-30°C
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



