EC Medium, CRITERION™, Hardy Diagnostics
2 kg, Bucket
Supplier: Hardy Diagnostics
Supplier #: C5692
EC Medium, CRITERION™, Hardy Diagnostics
Catalog # 89405-424
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Jump to:
Product Details & Documents
CRITERION™ EC Medium is recommended for the detection of coliforms at 35 degrees C
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
and Escherichia coli and fecal coliforms at 45.5 degrees C. Dehydratedculture media with engineered container.
Certifications: BSE Free
Documents
Specifications
Specifications
- Pack type:Bucket
- Size:2 kg
- Cat. No.:89405-424
- Storage Temperature:–2-30°C
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



