Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Stir Bar, Triangular, PTFE, Dynalon

51 mm (2"), 9.5 mm (3/8 "), PTFE, 0.02375 lbs.
Catalog # 30621-016
Supplier:   Dynalon
 :  302836
Stir Bar, Triangular, PTFE, Dynalon
Catalog # 30621-016
Supplier:  Dynalon
Stir Bar, Triangular, PTFE, Dynalon
Stir Bar, Triangular, PTFE, Dynalon

51 mm (2"), 9.5 mm (3/8 "), PTFE, 0.02375 lbs.

Price
$193.48
Log in for availability
Case of 12
Product Details & Documents

Triangular shape provides the best possible spinning action and most efficient mixing of all stir bars

Powerful permanent Alnico V magnetic cores. PTFE offers excellent chemical resistance. Always store in pairs to maintain magnetic strength.

Specifications
  • Coating:
    PTFE
  • Ø:
    9.5 mm (3/8 ")
  • Length:
    51 mm (2")
  • Weight:
    0.02375 lbs.