Orders LinkContactUsLinkComponent
mini-BUCK™ Primary Flow Calibrator, Electron Microscopy Sciences
Low Flow Calibrator, 120 V, 0.1 - 300 cc/min
Catalog # 102681-152
 :  65240-60
mini-BUCK™ Primary Flow Calibrator, Electron Microscopy Sciences
Catalog # 102681-152
mini-BUCK™ Primary Flow Calibrator, Electron Microscopy Sciences
mini-BUCK™ Primary Flow Calibrator, Electron Microscopy Sciences
Low Flow Calibrator, 120 V, 0.1 - 300 cc/min
Price
$2,618.15
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The mini-BUCK™ Primary Flow Calibrator is the industry standard for day-to-day flow verification and calibration of your personal and environmental sampling pumps.
  • No more lugging oversized calibration panels
  • No more rotameter inaccuracies
It is also suitable for verifying rotameters, setting instrument panels, and calibrating laboratory instruments. The mini-BUCK™ is accurate at any attitude and can be used for flow measurements with sampling media.
Specifications
  • Flow rate:
    0.1 - 300 cc/min
  • Description:
    Low Flow Calibrator, 120 V