Calibration Micrometer, Electron Microscopy Sciences
This ideal standard provides accurate calibration of any eyepiece, reticules, filar micrometers, or electronic measuring instrument
Features a series of ʽH’ shaped fiducial images in a range of sizes from 0.1 mm through 20 mm, useful for calibration range from 1X to 1000X power.
- Description:Model ACM-1, with 11 pts Calibrated