Stage Micrometer Model SM-5, Electron Microscopy Sciences
Product Details & Documents
Linear Reference Scales are available in Chrome-on-Glass (G) and Chrome-on-Opal glass (O) or Chrome-on Quartz (Q)
Positive image are mostly clear or white.
Ordering information: The Opal and Quartz are offered as a special only and must be requested.
Packaging: All EMS micrometers come in a sturdy wooden box.
Specifications
- Description:Stage Micrometer SM-5, Chrome on glass