Orders LinkContactUsLinkComponent
Stage Micrometer Model SM-4, Electron Microscopy Sciences
Stage Micrometer Model SM-4, Electron Microscopy Sciences
Stage Micrometer SM-4, Chrome on glass
Catalog # 102680-930
Supplier #:  60210-4PG
Stage Micrometer Model SM-4, Electron Microscopy Sciences
Catalog # 102680-930
Stage Micrometer Model SM-4, Electron Microscopy Sciences
Stage Micrometer Model SM-4, Electron Microscopy Sciences
Stage Micrometer SM-4, Chrome on glass
Price
$2,986.29
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Linear Reference Scales are available in Chrome-on-Glass (G) and Chrome-on-Opal glass (O) or Chrome-on Quartz (Q)
Positive image are mostly clear or white.
Ordering information: The Opal and Quartz are offered as a special only and must be requested.
Packaging: All EMS micrometers come in a sturdy wooden box.
Specifications
  • Description:
    Stage Micrometer SM-4, Chrome on glass