Stage Micrometer Model SM-1, Electron Microscopy Sciences
Product Details & Documents
Linear Reference Scales are available in Chrome-on-Glass (G) and Chrome-on-Opal glass (O) or Chrome-on Quartz (Q)
Positive image are mostly clear or white
: The Opal and Quartz are offered as a special only and must be requested.
: All EMS micrometers come in a sturdy wooden box.
Specifications
- Description:Stage Micrometer, SM-1 Chrome on Glass with Calibration