Orders LinkContactUsLinkComponent
MicroSaw, Diamond Saw, Electron Microscopy Sciences
MicroSaw, Diamond Saw, Electron Microscopy Sciences
MicroSaw, Model MS 3
Catalog # 102680-882
Supplier #:  55150-MS3
MicroSaw, Diamond Saw, Electron Microscopy Sciences
Catalog # 102680-882
MicroSaw, Diamond Saw, Electron Microscopy Sciences
MicroSaw, Diamond Saw, Electron Microscopy Sciences
MicroSaw, Model MS 3
Price
$6,407.76
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Operation: The Model MS 3 MicroSaw is driven by an electric motor through a helical transmission
This circular diamond saw is for precision cutting under stereo microscope. MicroSaw is a small, versatile diamond wheel saw, to be mounted under a standard laboratory microscope. The precision offered by such an optical alignment makes MicroSaw ideal for cutting and sectioning required during the TEM specimen preparation. The compact design and easy operation, MicroSaw allows the user cut almost any solid material with precisely and reproducibly prior to thinning for TEM. Thin ceramic, semiconductor material, hard metals can be further sectioned with 0.01 mm accuracy.
Operation: The Model MS 3 MicroSaw is driven by an electric motor through a helical transmission. The 12V DC is provided by the power supply subunit. The speed at which the wheel turns is continuously adjustable. Adjustment of the specimen position, arm contact point, arm tension and the down-stop control mechanism is possible. Two adjustment screws at the bottom of the instrument allow positioning of the stand, and leveling the MicroSaw underneath of the stereomicroscopy for precision sectioning.
System Configuration:
Switching power supply 100 – 240 VAC, 50/60 Hz
MicroSaw Model MS 3 with 1 diamond wheel
System Configuration:
Switching power supply 100 – 240 VAC, 50/60 Hz
MicroSaw Model MS 3 with 1 diamond wheel
The 12V DC is provided by the power supply subunit. The speed at which the wheel turns is continuously adjustable. Adjustment of the specimen position, arm contact point, arm tension and the down-stop control mechanism is possible. Two adjustment screws at the bottom of the instrument allow positioning of the stand, and leveling the MicroSaw underneath of the stereomicroscopy for precision sectioning.
Specifications
  • Description:
    MicroSaw, Model MS 3