Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
MIXED ACID ETCHANT 2:1:0-ENTEGRIS PFA250
Catalog # 89400-824
Supplier:
KMG
SDS
Expand image
MIXED ACID ETCHANT 2:1:0-ENTEGRIS PFA250
Catalog # 89400-824
Supplier:
KMG
Supplier Number:
200507
SDS
Related Information
Documentation
SDS
ghs_200507
Additional Product Details
VWR GHS Compliance Letter (US)
Documentation