Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BUFFERED OXIDE ETCH 20:1-POLY BOTTLE-17
Catalog # 89400-754
Supplier:
KMG
SDS
Expand image
BUFFERED OXIDE ETCH 20:1-POLY BOTTLE-17
Catalog # 89400-754
Supplier:
KMG
Supplier Number:
62055
SDS
Related Information
Documentation
SDS
ghs_62055
Additional Product Details
VWR GHS Compliance Letter (US)
Documentation