Orders LinkContactUsLinkComponent
Solid Caps, GL-Threads, Polypropylene, Chemglass
Solid Caps, GL-Threads, Polypropylene, Chemglass
Solid Caps, Gray, GL45
Catalog # 30623-016
Supplier:   Chemglass
Supplier #:  CLS-1480-14
Solid Caps, GL-Threads, Polypropylene, Chemglass
Catalog # 30623-016
Supplier:  Chemglass
Solid Caps, GL-Threads, Polypropylene, Chemglass
Solid Caps, GL-Threads, Polypropylene, Chemglass
Solid Caps, Gray, GL45
Price
$70.10
Log in for availability
Pack of 10
Product Details & Documents
These solid polypropylene caps can be autoclaved in steam to 140 °C.
Specifications
  • Color:
    Gray
  • Description:
    Solid Caps
  • Thread size:
    GL45