Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Trays, Rectangular, Polypropylene, Dynalon

Trays, Rectangular, Polypropylene, Dynalon

10L x 15W x 3H"
Catalog # 30620-520
Supplier:   Dynalon
Supplier #:  107304
Trays, Rectangular, Polypropylene, Dynalon
Catalog # 30620-520
Supplier:  Dynalon
Trays, Rectangular, Polypropylene, Dynalon
Trays, Rectangular, Polypropylene, Dynalon

10L x 15W x 3H"

Price
$176.30
Log in for availability
Each
Product Details & Documents

Heavy trays fabricated with 3/16" thick autoclavable Polypropylene

All welded construction. Trays can be customized with spigots and covers which are fabricated to order.

Ordering information: Additional sizes and materials are available upon request.

Specifications
  • Interior dimensions:
    10L x 15W x 3H"
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.