Stirring Rod, Solid, Dynalon
8 mm, 200 mm, 0.02125 lbs.
Catalog # 30620-512
Supplier: Dynalon
: 303334-0002
Jump to:
Product Details & Documents
Pure PTFE solid stirring rod with flat tapered ends
Inert and will not scratch, the rod can be used up to 280°C. PTFE has excellent chemical resistance and is autoclavable.
Specifications
- Ø:8 mm
- Length:200 mm
- Weight:0.02125 lbs.
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



