Stirring Rod, Solid, Dynalon
8 mm, 300 mm, 0.04625 lbs.
Catalog # 30620-506
Supplier: Dynalon
: 303334-0004
Jump to:
Product Details & Documents
Pure PTFE solid stirring rod with flat tapered ends
Inert and will not scratch, the rod can be used up to 280°C. PTFE has excellent chemical resistance and is autoclavable.
Specifications
- Ø:8 mm
- Length:300 mm
- Weight:0.04625 lbs.



