Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Stirring Rod, Solid, Dynalon

8 mm, 250 mm, 0.03375 lbs.
Catalog # 30620-504
Supplier:   Dynalon
 :  303334-0003
Stirring Rod, Solid, Dynalon
Catalog # 30620-504
Supplier:  Dynalon
Stirring Rod, Solid, Dynalon
Stirring Rod, Solid, Dynalon

8 mm, 250 mm, 0.03375 lbs.

Price
$183.51
Log in for availability
Case of 6
Product Details & Documents

Pure PTFE solid stirring rod with flat tapered ends

Inert and will not scratch, the rod can be used up to 280°C. PTFE has excellent chemical resistance and is autoclavable.

Specifications
  • Ø:
    8 mm
  • Length:
    250 mm
  • Weight:
    0.03375 lbs.