Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Hydrometer Racks, Polypropylene, Dynalon

Tall PP rack, L×W×H 7.8×8.5×10", plate height 7"
Catalog # 30620-398
Supplier:   Dynalon
 :  103215-0000
Hydrometer Racks, Polypropylene, Dynalon
Catalog # 30620-398
Supplier:  Dynalon
Hydrometer Racks, Polypropylene, Dynalon
Hydrometer Racks, Polypropylene, Dynalon

Tall PP rack, L×W×H 7.8×8.5×10", plate height 7"

Price
$182.13
Log in for availability
Each
Product Details & Documents

The 18-place polypropylene racks are specially designed to safeguard glass hydrometers.

  • The three tiered rack with handles allows easy transport and storage of hydrometers
  • Top two plates have 1" diameter holes while the bottom plate has wells which taper from 1" to ⅜"

Tall 7" and short 5⅜" version available

 : Additional sizes and materials are available upon request.

Specifications
  • Description:
    Tall PP rack, L×W×H 7.8×8.5×10", plate height 7"