Calcium hydroxide, Technical Grade
1 Kg
Calcium hydroxide, Technical Grade
Catalog # 89399-874
CAS Number: 1305-62-0
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Product Details & Documents
Synonyms: Slaked lime, Calcium dihydroxide
- CAS Number:1305-62-0
- Formula:Ca(OH)₂
- MDL Number:MFCD00010901
- Molecular Weight:74.09 g/mol
- Merck Index:13,01675
- Melting Pt:550 °C
- Density:2.24 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-137-3
- UN:3262
Specifications
Specification Test Results
- Calcium Carbonate {CaCO3}:< 15%
- Calcium Hydroxide {Ca(OH)2}:> 83 %
Specifications
- Size:1 Kg
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



