Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Calcium hydroxide, Technical Grade

Calcium hydroxide, Technical Grade

500 g
Catalog # 89399-872
Corrosive
Irritant
Supplier:   VWR
Supplier #:  N883-500G
Calcium hydroxide, Technical Grade
Catalog # 89399-872
Supplier:  VWR
CAS Number:  1305-62-0
Calcium hydroxide, Technical Grade
Calcium hydroxide, Technical Grade

500 g

CAS Number:  1305-62-0
Price
$90.50
Log in for availability
Each (500g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Slaked lime, Calcium dihydroxide
  • CAS Number:1305-62-0
  • Formula:Ca(OH)₂
  • MDL Number:MFCD00010901
  • Molecular Weight:74.09 g/mol
  • Merck Index:13,01675
  • Melting Pt:550 °C
  • Density:2.24 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-137-3
  • UN:3262
Specifications

Specification Test Results

  • Calcium Carbonate {CaCO3}:
    < 15%
  • Calcium Hydroxide {Ca(OH)2}:
    > 83 %

Specifications

  • Size:
    500 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.