Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

MEGA FlexTubes, IBI Scientific

MEGA FlexTube Kit, 10 ml, 1 K
Catalog # 89369-600
Supplier:   IBI Scientific
 :  IB48310
MEGA FlexTubes, IBI Scientific
Catalog # 89369-600
Supplier:  IBI Scientific
MEGA FlexTubes, IBI Scientific
MEGA FlexTubes, IBI Scientific

MEGA FlexTube Kit, 10 ml, 1 K

Price
$198.52
Log in for availability
Pack of 5
Product Details & Documents

IBI MEGA FlexTubes are typically used for dialysis or buffer exchange with volumes of 3 to 20 ml.

The FlexTube membrane is ultra-clean, sulfur- and heavy metal-free, and EDTA treated.

IBI FlexTubes allow rapid, secure, and simple loading and recovery of samples. IBI FlexTubes can also be used for applications such as the preparation of protein samples for MALDI-MS, sample concentration, tissue culture extraction purification, removal of salts, surfactants, solvents, and detergents, as well as peptide dialysis and virus-particle purification. The performance of IBI MEGA FlexTubes are regularly monitored and tested by simple dialysis and assays commonly used for proteins and/or nucleic acids.

Specifications
  • Volume:
    10 ml
  • Description:
    MEGA FlexTube Kit
  • MWCO:
    1 K