Orders LinkContactUsLinkComponent
Corning® Polypropylene Erlenmeyer Flasks, Corning
Corning® Polypropylene Erlenmeyer Flasks, Corning
125 ml, 20 ml, GL-40
Catalog # 30617-088
Supplier #:  4985P-100
Corning® Polypropylene Erlenmeyer Flasks, Corning
Catalog # 30617-088
Supplier:  
Corning® Polypropylene Erlenmeyer Flasks, Corning
Corning® Polypropylene Erlenmeyer Flasks, Corning
125 ml, 20 ml, GL-40
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
FLASK ERLMYR PP NM 250ML CS6 Catalog #30617-090
Product 30617-090 is similar to 30617-088 but manufacturer and specifications may vary.
ERLENMEYER FLASK VITLAB PP CAP 75 ML PK6 Catalog #76313-834
Product 76313-834 is similar to 30617-088 but manufacturer and specifications may vary.
Product Details & Documents
PP, translucent.
  • Very good chemical resistance
  • Very good temperature resistance
With blue, easy to read graduations. Autoclavable. The flasks feature a stopper bed and can also be sealed with standard ground joint polypropylene stoppers.  
Specifications
  • Graduation Interval:
    20 ml
  • Capacity:
    125 ml
  • Thread:
    GL-40