Orders LinkContactUsLinkComponent
Corning® Polypropylene Erlenmeyer Flasks, Corning
Corning® Polypropylene Erlenmeyer Flasks, Corning
50 ml, 10 ml, GL-40
Catalog # 30617-086
Supplier #:  4985P-50
Corning® Polypropylene Erlenmeyer Flasks, Corning
Catalog # 30617-086
Supplier:  
Corning® Polypropylene Erlenmeyer Flasks, Corning
Corning® Polypropylene Erlenmeyer Flasks, Corning
50 ml, 10 ml, GL-40
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
FLASK ERLENMEYER 50 ML PP PK6 Catalog #83008-938
Product 83008-938 is similar to 30617-086 but manufacturer and specifications may vary.
Product Details & Documents
PP, translucent.
  • Very good chemical resistance
  • Very good temperature resistance
With blue, easy to read graduations. Autoclavable. The flasks feature a stopper bed and can also be sealed with standard ground joint polypropylene stoppers.  
Specifications
  • Graduation Interval:
    10 ml
  • Capacity:
    50 ml
  • Thread:
    GL-40