Molybdate 3 Reagent Solution, Hach
Molybdate 3 Reagent Solution, 100 mL Marked Dropping Bottle, Heteropoly blue, 100
Supplier: Hach
: 199532
Molybdate 3 Reagent Solution, Hach
Catalog # 89231-256
Supplier: Hach
Product available on GSA Advantage®
CAS Number:
Jump to:
Product Details & Documents
For low and ultra-low range silica determination by the Heteropoly Blue method
Hach methods 8282 and 8186.
: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.
Specifications
Specifications
- Chemical method:Heteropoly blue
- Description:Molybdate 3 Reagent Solution
- Size:100 mL Marked Dropping Bottle
- No. of tests:100
- Cat. No.:89231-256
- Method:Heteropoly Blue