Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Colchicine

Colchicine

Vial, 1 g
Catalog # 102515-852
Toxic
Health
Marketsource
Supplier:   Adipogen
Supplier #:  AG-CN2-0048-G001
Colchicine
Catalog # 102515-852
Supplier:  Adipogen
Marketsource
CAS Number:  64-86-8
Colchicine

Vial, 1 g

CAS Number:  64-86-8
Price
$166.15
Log in for availability
Each (1g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Anti-cancer compound. Microtubule assembly inhibitor. Depolymerizes microtubules and limits microtubule formation (inactivates spindle fibre formation). Inhibits mitosis during cell division at metaphase by inhibiting spindle formation. Anti-inflammatory compound. Suppresses monosodium urate crystal-induced NLRP3/NALP3 inflammasome-driven caspase-1 activation, IL-1beta processing and release, and L-selectin expression on neutrophils at micromolar concentrations. Blocks the release of a crystal-derived chemotactic factor from neutrophil lysosomes, blocks neutrophil adhesion to, and inhibits monosodium urate crystal-induced production of superoxide anions from neutrophils at nanomolar concentrations. Drug used in treatment of gout, familial Mediterranean fever, pericarditis and Behect's disease. Investigated for its anti-cancer activity. It has a narrow therapeutic index with no clear-cut distinction between nontoxic, toxic and lethal doses, causing substantial confusion among clinicians. Apoptosis inducer in a variety of normal and tumor cell lines. Inhibitor of autophagosome-lysosome fusion. Inhibits acetylated a-tubulin mediated dynein dependent transport of mitochondria and subsequent apposition of ASC on mitochondria to NLRP3 on the endoplasmic reticulum in vitro and in vivo.

Synonyms:  7α-H-Colchicine, (S)-(-)-Colchicine
  • CAS Number:64-86-8
  • Formula:C₂₂H₂₅NO₆
  • MDL Number:MFCD00078484
  • Molecular Weight:399.44 g/mol
  • Storage Temperature:Freezer
  • UN:3462
  • ADR:6.1,III
Specifications

Specification Test Results

  • Source/Host:
    Isolated from Colchicum autumnale.
  • Purity:
    >98% (HPLC)
  • Appearance/Color:
    White to off-white powder.
  • Solubility:
    Soluble in 100% ethanol (>100mg/ml), DMSO (>100mg/ml), water or DMF.

Specifications

  • Pack type:
    Vial
  • Size:
    1 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.