Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Pentafluoropropionic anhydride ≥98%

Pentafluoropropionic anhydride ≥98%

25 g
Catalog # 101778-884
Marketsource
Supplier:   Matrix Scientific
Supplier #:  006130-25G
Pentafluoropropionic anhydride ≥98%
Catalog # 101778-884
Supplier:  Matrix Scientific
Marketsource
CAS Number:  356-42-3
Pentafluoropropionic anhydride ≥98%

25 g

CAS Number:  356-42-3
Price
$184.62
Log in for availability
Each (25g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

TCA listed: T
Hazard: Corrosive/Moisture Sensitive/Refrigerate

Synonyms:  Perfluoropropanoic anhydride, Pentafluoropropanoic anhydride, PFPA
  • CAS Number:356-42-3
  • Formula:C₆F₁₀O₃
  • MDL Number:MFCD00000429
  • Molecular Weight:310.05
  • Boiling Pt:69…70 °C (735 mmHg)
  • Density:1.571
  • UN:3265
  • ADR:8,II
  •  :Density: 1.571 g/cm³ (25 °C)

Documents

Specifications

Specification Test Results

  • Boiling point:
    69-70°/735
  • Density:
    1.571

Specifications

  • Size:
    25 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.