Orders LinkContactUsLinkComponent
ProtoScript II Reverse Transcriptase, New England Biolabs
ProtoScript II Reverse Transcriptase, New England Biolabs
ProtoScript II Reverse Transcriptase, 4,000 units
Catalog # 102500-080
Supplier #:  M0368S
ProtoScript II Reverse Transcriptase, New England Biolabs
Catalog # 102500-080
CAS Number:  
ProtoScript II Reverse Transcriptase, New England Biolabs
ProtoScript II Reverse Transcriptase, New England Biolabs
ProtoScript II Reverse Transcriptase, 4,000 units
CAS Number:  
Price
$112.00
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
ProtoScript II Reverse Transcriptase is a recombinant M-MuLV reverse transcriptase with reduced RNase H activity and increased thermostability
It can be used to synthesize first strand cDNA at higher temperatures than the wild type M-MuLV. The enzyme is active up to 50°C, providing higher specificity, higher yield of cDNA and more full length cDNA product, up to 12 kb in length. This product was formerly known as M-MuLV Reverse Transcriptase RNase H–.
Specifications
Specifications
  • Description:
    ProtoScript II Reverse Transcriptase
  • Size:
    4,000 units