Orders LinkContactUsLinkComponent
GL Series Stereo Microscopes, Electron Microscopy Sciences
Stereo Boom Stand Trinocular 7:1 Zoom Ratio
Catalog # 102101-330
GL Series Stereo Microscopes, Electron Microscopy Sciences
Catalog # 102101-330
Supplier:  
GL Series Stereo Microscopes, Electron Microscopy Sciences
GL Series Stereo Microscopes, Electron Microscopy Sciences
Stereo Boom Stand Trinocular 7:1 Zoom Ratio
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The remarkably clear, non-distorted 3D image resolved by the GL Series optics is a result of advanced technologies in optical design. The GL Series Greenough design produces lifelike images in a large field of view and presents upright or erect images with astounding depth perception.
  • No need to refocus when zooming magnification, the GL Series is parfocal and its continuous zoom is available with detentes at 0.5X increments for precise measurements
  • Wide interpupillary adjustment (45 - 75mm) and diopters built in to the eyepieces allow virtually all individuals to work in comfort
  • Minimize fatigue - side mounted zoom controls and low positioned focus control with tension adjustment reduce stress and let the user rest in a natural position
  • High contrast, flat images are now cost effective and EMS’s 5 year limited warranty protects your investment
  • 3 Zoom Ranges
A long (100 mm) working distance facilitates observation. The Illuminated stand allows observation of both opaque and transparent specimens because of individual controls for reflected and transmitted light. While reflected or incident light is a 15 W Halogen with electronic dimmer, the transmitted light can be either a 15W Halogen with dimmer or a “cool” 5 W fluorescent bulb for heat sensitive samples. Superb resolution meets demands in semi-conductor/high tech industries, medical device assembly and life science research. Long duration tasks are less fatiguing with accessible controls. Individual lighting controls provide ideal conditions for photographing images in a fully illuminated field.
The GL7 Optical Body has brilliance, clarity, parfocality, flatness and depth of field that characterize this hard working model. Its wide magnification range, (7:1 or 0.65x to 4.5x) zooms in on your specimen without distortion, delivering a resolved image true to character. The detents at 0.5X allow exactly repeatable magnifications for documentation. The GL6 Optical Body is designed for those demanding applications where you are challenged with new and different specimens requiring absolute clarity at all powers, this model achieves high magnification (1.0x to 6.5x, 6:1 ratio) at a low price. Going beyond the standard for this class of microscope, total magnification ranges from 0.5x to 260x with optional auxiliary lenses and eyepieces. GL Series optical bodies and stands master Industrial environments.
The GL7 boasts a large field of view of 35 mm. The compact design/long working distance facilitates machine mounts. Large zoom range leads to accurate reproduction of the sample up to 260X. There is a wide range of imaging functions of fully integrated camera/software systems.
The Post and Boom stands are heavy duty and designed to provide ample support for many years. Allowing freedom of movement in multiple directions over the work area to facilitate inspection of large samples, their rugged structure minimizes vibration and tipping, even on a Trinocular head with camera attached.
Boom Stands feature heavy bases for stability and tilting/swiveling mounts to facilitate observation of even hard to get to areas. Standard Boom is economical and well-matched to applications where continuous horizontal movement is not required. Smooth Boom got its name from the smooth gliding motion produced by the ball bearings in the horizontal movement. Articulating Arm Boom features a counter balanced arm that moves vertically as well as horizontally, making it ideal for tight spaces.
Delivery information: All models include: a pair of 10x eye pieces, black/white reversible stage plate, dust cover, power cord and instruction manual.
Specifications
  • Description:
    Stereo Boom Stand Trinocular 7:1 Zoom Ratio