Orders LinkContactUsLinkComponent
Formvar Film on Hexagonal Grids, Electron Microscopy Sciences
Formvar Film on Hexagonal Grids, Electron Microscopy Sciences
Hexagonal Grid, Nickel, 5 - 6 nm, 200
Catalog # 102100-806
Supplier #:  FF200H-NI-25
Formvar Film on Hexagonal Grids, Electron Microscopy Sciences
Catalog # 102100-806
Formvar Film on Hexagonal Grids, Electron Microscopy Sciences
Formvar Film on Hexagonal Grids, Electron Microscopy Sciences
Hexagonal Grid, Nickel, 5 - 6 nm, 200
Price
$78.73
Log in for availability
Box of 25
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A thin film of pure formvar resin coats the shiny side of a Hexagonal grid.
  • Formvar coatings thickness of standard (10 nm), ultra-thin (5 to 6 nm), thick (15 to 20 nm), or extra thick (25 to 50 nm)
  • Select from copper, nickel, and gold grids in 100, 200, 300, and 400 mesh sizes
Optically checked followed by batch testing in the EM.
 : Packed in grid storage box.
Specifications
  • Description:
    Hexagonal Grid
  • Material:
    Nickel
  • Thickness:
    5 - 6 nm
  • Mesh:
    200