Formvar Film on Hexagonal Grids, Electron Microscopy Sciences
A thin film of pure formvar resin coats the shiny side of a Hexagonal grid.
- Formvar coatings thickness of standard (10 nm), ultra-thin (5 to 6 nm), thick (15 to 20 nm), or extra thick (25 to 50 nm)
- Select from copper, nickel, and gold grids in 100, 200, 300, and 400 mesh sizes
Optically checked followed by batch testing in the EM.
: Packed in grid storage box.
- Description:Hexagonal Grid
- Material:Gold
- Thickness:5 - 6 nm
- Mesh:100