Copper, sputtering target sputter coater cathodes
Product Details & Documents
EMS (ELECTRON MICROSCOPY SCIENCES) precious metal sputtering targets are made from high purity metals, starting 99.99% with most standard disc sizes, for use on most sputtering units from many manufacturers: Emitech, Emscope, Bio-Rad, Polaron, Edwards, Balzers, Plasma Sciences, Technics Hummers, Denton, Cressington, and much more.
EMS targets come in a standard thickness of 0.1 mm (100µm). Other thickness, ranging from 0.05 to 6mm, are available upon request. Targets are avaible mounted or unmounted whichever you prefer. For mounting please always state the make and the model of the sputtering unit so we can tend to it for you. Targets in other materials such as Chromium, Iridium etc. are available upon request. We also can manufacture targets with diameters from 4 mm to 304.8 mm (12"). Call for pricing.
- CAS Number:7440-50-8
- Formula:Cu
- MDL Number:MFCD00010965
- Molecular Weight:63.55 g/mol
Specifications
Specifications
- Size:57 mm
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



