Orders LinkContactUsLinkComponent
Copper, sputtering target sputter coater cathodes
20 mm
  102099-402
 :  91006-CU
Copper, sputtering target sputter coater cathodes
  102099-402
 :  7440-50-8
Copper, sputtering target sputter coater cathodes
Copper, sputtering target sputter coater cathodes
20 mm
 :  7440-50-8
$193.81
Each
EMS (ELECTRON MICROSCOPY SCIENCES) precious metal sputtering targets are made from high purity metals, starting 99.99% with most standard disc sizes, for use on most sputtering units from many manufacturers: Emitech, Emscope, Bio-Rad, Polaron, Edwards, Balzers, Plasma Sciences, Technics Hummers, Denton, Cressington, and much more.
EMS targets come in a standard thickness of 0.1 mm (100µm). Other thickness, ranging from 0.05 to 6mm, are available upon request. Targets are avaible mounted or unmounted whichever you prefer. For mounting please always state the make and the model of the sputtering unit so we can tend to it for you. Targets in other materials such as Chromium, Iridium etc. are available upon request. We also can manufacture targets with diameters from 4 mm to 304.8 mm (12"). Call for pricing.
  •  :7440-50-8
  •  :Cu
  •  :MFCD00010965
  •  :63.55 g/mol
  • Size:
    20 mm