Light Pipe Scintillators, Electron Microscopy Sciences
The light pipe is the connection between the scintillator and the phosphomultiplier tube (PMT). It transfers the photons produced by the scintillator for detection by the PMT.
- Scintillator material coated directly on the light pipe which improves the signal
- Re-coating is typically needed twice per year
Light Pipes are available in optical quality quartz and optical grade acrylic. Cambridge reduced-tip models are only available in acrylic.
: Includes one Re-coating Certificate. High voltage wire and corona ring are included, JEOL pipes include a flange.
- Description:Cambridge Light Pipe Scintillator, Acrylic, Reduced Tip, any