Orders LinkContactUsLinkComponent
Micro Scissors, Rubis, Electron Microscopy Sciences
Micro Scissors, Rubis, Electron Microscopy Sciences
Style Micro Scissors Set, —
Catalog # 102098-974
Supplier #:  78180-03
Micro Scissors, Rubis, Electron Microscopy Sciences
Catalog # 102098-974
Micro Scissors, Rubis, Electron Microscopy Sciences
Micro Scissors, Rubis, Electron Microscopy Sciences
Style Micro Scissors Set, —
Price
$77.84
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Micro scissors manufactured from hardened, anti-acid, stainless, surgical steel (CA)
Rubis Micro Scissors Set includes one light duty Rubis Colibri fine scissors, one medium duty Rubis Classic medium fine scissors, and one heavy duty Rubis Sauro medium strong scissors. Strong, slightly curved easily cut through small bones and cartilages.
Handcraft with precision sharpened blades that provide a perfect cut. The Rubis Twist® grip scissors offer ergonomically, comfort and control.
Specifications
  • Description:
    Style Micro Scissors Set
  • Blade length:
Some Products May Appear Restricted