Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Labeling Tape, Electron Microscopy Sciences

Labeling Tape, Electron Microscopy Sciences

Blue, 0.75" x 2160"
Catalog # 102098-766
Supplier #:  77628-B
Labeling Tape, Electron Microscopy Sciences
Catalog # 102098-766
Labeling Tape, Electron Microscopy Sciences
Labeling Tape, Electron Microscopy Sciences

Blue, 0.75" x 2160"

Price
$58.95
Log in for availability
Roll of 1
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Use pen, pencil or marker to mark. Leaves no adhesive residue when peeled off.

Water, oil, and acid resistant. Withstands cold (−23 °C) and hot (121 °C) temperatures.

Specifications
  • Color:
    Blue
  • Size:
    0.75" x 2160"

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.