Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Write Protect Tape, Electron Microscopy Sciences

Write Protect Tape, Electron Microscopy Sciences

Write Protect Tape, 41 x 16 mm
Catalog # 102098-754
Supplier #:  77626-20
Write Protect Tape, Electron Microscopy Sciences
Catalog # 102098-754
Write Protect Tape, Electron Microscopy Sciences
Write Protect Tape, Electron Microscopy Sciences

Write Protect Tape, 41 x 16 mm

Price
$55.28
Log in for availability
Roll of 1,000
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Write Protect is a clear adhesive tape designed to cover and protect writing or printing on labels, on the surface of microtubes and on other laboratory containers.

  • Will not dry out, peel or fall off
  • Withstand temperatures ranging from –196 to 150 °C

The clear tape comes in a handy dispenser and will adhere to all plastics, glass, metals and label materials. The tape is provided in perforated, pull-off segments for 0.5 and 1.5 mL tubes.

Specifications
  • Description:
    Write Protect Tape
  • Dimensions:
    41 x 16 mm

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.